KIMBLE® Jacketed Coil Type Reflux Condensers, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
457270-2425 457250-0225
KT457270-2425CS 763.87 USD
KT457270-2425 KT457250-0225
KIMBLE® Jacketed Coil Type Reflux Condensers, DWK Life Sciences
Condensers
Condenser with water-cooled outer jacket to increase condensation capability.
  • Full length 24/40 joint
  • Reflux condenser with an internal coil-type cold finger
  • Vapors spiraling upward are condensed on the inner wall of the jacket and on the outer surface of the coil
  • Manufactured from 33 expansion, low extractable borosilicate glass
These condenser has a standard taper outer joint at the top, a standard taper inner drip joint at the bottom and two hose connectors at the top for water inlet and outlet. It has a straight outlet at the top.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.