5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
5-Ethyltio-1H-Tetrazole
BDH0604-2.5L
BDH0604-4L
BDH0604-56L
BDH0604-2.5LCS
5374.28
USD
BDH0604-2.5L
BDH0604-4L
BDH0604-56L
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
5-(Ethylthio)-1H-tetrazole (ETT)
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.
- Custom Blends and Bulk Sizes available upon request.
Formula:
C₃H₆N₄S MW: 130.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00068733 CAS Number: 89797-68-2 UN: 1648 ADR: 4 II |
Specification Test Results
Description (Clear liquid free of suspended matter) | Pass |
5-Ethythio-1H-tetrazole concentration | 0.48 - 0.52 M |
Color (at time of packaging) | <30 APHA |
Water | Max 30 ppm |
Acetonitrile (component purity) | 99.9% |
5-Ethythio-1H-tetrazole (component purity) | 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...