5-Benzylmercaptotetrazole (BMT, BTT) 0.30M in anhydrous acetonitrile/1-methylimidazole (NMI) [99.5:0.5] activator reagent, for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: BTT5-(Benzylthio)-1H-tetrazoleBMT

BDH0603-45 AB3961-014 AB3961-001
BDH0603-450MLCS 1607.4 USD
BDH0603-450ML BDH0603-4L BDH0603-2.5L
5-Benzylmercaptotetrazole (BMT, BTT) 0.30M in anhydrous acetonitrile/1-methylimidazole (NMI) [99.5:0.5] activator reagent, for biosynthesis, VWR Chemicals BDH®
5-(Benzylthio)-1H-tetrazole

BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.


  • Custom Blends and Bulk Sizes available upon request.

Formula: C₈H₈N₄S
MW: 192.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068731
CAS Number: 21871-47-6
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Description (Clear liquid free of suspended matter) Pass
Benzylthiotetrazole concentration 0.285 - 0.315 M
Color (at time of packaging) <100 APHA
Water Max 30 ppm
Acetonitrile (component purity) 99.9%
Benzylthiotetrazole (component purity) 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR