5-Benzylmercaptotetrazole (BMT, BTT) 0.20M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
BTT
, 5-(Benzylthio)-1H-tetrazole
, BMT
AB3964-112
AB3964-014
AB3964-157
AB3964-032
AB3964-051
AB3964-001
BDH0601-2.5LCS
3667.67
USD
BDH0601-2.5L
BDH0601-4L
BDH0601-200L
BDH0601-450ML
BDH0601-56L
BDH0601-20L
5-Benzylmercaptotetrazole (BMT, BTT) 0.20M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
5-(Benzylthio)-1H-tetrazole
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.
- Custom Blends and Bulk Sizes available upon request.
Formula:
C₈H₈N₄S MW: 192.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00068731 CAS Number: 21871-47-6 UN: 1648 ADR: 3,II |
Specification Test Results
Description (Clear Liquid free of suspended matter) | Pass |
Benzylthiotetrazole Concentration | 0.19 - 0.21 M |
Color | <100 APHA |
Water | Max 20 ppm |
Acetonitrile (component purity) | 99.9% |
Benzylthiotetrazole (component purity) | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...