Sulfuric acid 2.5 mol/l (5 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

4.80364.1000 4.80364.9025
EM4.80364.1000CS 218 USD
EM4.80364.1000 EM4.80364.9025
Sulfuric acid 2.5 mol/l (5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.15 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR