Sulfuric acid 0.05 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ100
1.09074.1000
1.09074.4000
1.09074.9010
EM1.09074.1000CS
194
USD
EM1.09074.1000
EM1.09074.4000
EM1.09074.9010
Sulfuric acid 0.05 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid
Formula:
H₂SO₄ Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 3264 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...