InTemp® Bluetooth® Temperature with Glycol Data Loggers
Supplier: ONSET COMPUTER CORPORATION SE
Bluetooth®
CX402-VFC430 CX402-VFC230 CX402-VFC205 CX402-VFC405 CX402-VFC215 CX402-VFC415
470323-342EA 223.69 USD
470323-342 470323-340 470323-322 470323-344 470323-338 470323-336
InTemp® Bluetooth® Temperature with Glycol Data Loggers
Data Loggers
InTemp ® CX402 (with Glycol Bottle) data loggers monitor temperatures in refrigerators and freezers for clinics participating in the CDC Vaccines for Children program as well as hospitals, clinics, and WHO sites.
  • Complies with all CDC and Vaccine for Children program requirements
  • Easy-to-read display with current and daily min/max temperatures
  • Bluetooth® transmission of data and alarm states
  • Real-time alerts and automatic data downloads with optional CX Gateway device
  • Logger setup and download of data in seconds – without a PC or cable
  • ±0.5 °C accuracy for reliable temperature readings
  • Battery-powered to guard against local power loss
  • Several glycol bottle sizes with probe and cable-length options
  • Includes three year ISO 17025 compliant calibration
The loggers communicate wirelessly via Bluetooth® Low Energy to mobile devices. Using the InTemp app, you can easily view data, check logger status, set alarms, and create and share secure PDF reports for streamlined reporting and regulatory compliance. All loggers feature a built-in LCD display to view current and daily min and max temperatures, advanced audible and visual alarm capabilities for notification of temperature excursions, an internal sensor for ambient temperature monitoring, and a three year NIST Certification of Calibration (Probe Only).
Models are available with a two or three meter external temperature probe attached to a Glycol bottle used to insulate the probe and delay thermal response times.