Anti-denatured collagen type-I (RGD motif) Mouse Monoclonal Antibody [clone: XL313]

Supplier: Bio X Cell
BE0324-25MG BE0324-100MG BE0324-5MG BE0324-50MG BE0324-1MG
103778-458EA 3920 USD
103778-458 103778-460 103778-462 103778-464 103778-466 76554-438 76554-564 76555-582 76556-014 76556-836
Anti-denatured collagen type-I (RGD motif) Mouse Monoclonal Antibody [clone: XL313]
Antibodies
Type: Primary
Antigen: denatured collagen type-I (RGD motif)
Clonality: Monoclonal
Clone: XL313
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: human, mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR