Flocculators, Velp
Supplier: VELP SCIENTIFIC INC.
F105A0108 F105A0109 F105A0117 F105A0111 F105A0112
76363-2398EA 2880.53 USD
76363-2398 76363-2400 76363-2402 76363-2404 76363-2406
Flocculators, Velp
Flocculators
A range of flocculator instruments to support technicians working in the environmental sector. The multiple place stirring provides more accurate data and reproducible results.
  • Timer function for FP4 and JLT units
  • Stainless steel stirring rods are adjustable in height
  • Strong metallic frame, epoxy coated for protection against chemical and mechanical corrosion
Portable unit FP4: The FP4 has four stirrers with reproducible stirring speeds allowing standard conditions to be met during on-site analysis. Central illumination of the four positions.