Paraformaldehyde 4% in 0.1 M phosphate buffered saline pH 7.4 fixative reagent
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
15735-85
15735-90
102091-888PK
91.59
USD
102091-888
102091-890
Paraformaldehyde 4% in 0.1 M phosphate buffered saline pH 7.4 fixative reagent
Formaldehyde
Formula:
CH₂O MW: 30.03 g/mol Boiling Pt: -19.3 Melting Pt: -92 Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 Merck Index: 13,04259 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...