Paraformaldehyde 4% in 0.2 M phosphate buffer pH 7.2 fixative reagent

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

15735-70 15735-80
102091-884PK 91.27 USD
102091-884 102091-886
Paraformaldehyde 4% in 0.2 M phosphate buffer pH 7.2 fixative reagent
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
Merck Index: 13,04259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240682c03df85e943acbb1a983fe71750db44cd&vid=96c37abe2cf4c6160dda3947fb05d9d23c32fa74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR