Paraformaldehyde 4% Sodium Cacodylate_0.1M pH 7.4 Fixative
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
15952-10S
15952-15-S
102091-940PK
106.54
USD
102091-940
102091-944
Paraformaldehyde 4% Sodium Cacodylate_0.1M pH 7.4 Fixative
Formaldehyde
Formula:
CH₂O MW: 30.03 g/mol Boiling Pt: -19.3 Melting Pt: -92 Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 Merck Index: 13,04259 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...