ICP Calibration Standard 7, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010725-100N
RCRV010725100NEA 485.76 USD
RCRV010725100N
ICP Calibration Standard 7, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
7 component standard in 5% hydrochloric acid
100 ppm: Hf, Ir, Sb, Sn, Ta, Ti, Zr
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility