Tin Standards, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010052-500N RV010083-500N RV010083-100N RV010052-100N
RCRV010052500NEA 121.6 USD
RCRV010052500N RCRV010083500N RCRV010083100N RCRV010052100N
Tin Standards, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Tin Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml