Scandium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010413-500N RV010413-250N RV010413-100N
RCRV010413250NEA 1556.91 USD
RCRV010413250N RCRV010413500N RCRV010413100N
Scandium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Scandium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml