Ruthenium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010308-250N
RV010308-500N
RV010410-100N
RCRV010308-50NEA
89.57
USD
RCRV010308-50N
RCRV010308100N
RCRV010308250N
RCRV010308500N
RCRV010410100N
RCRV010410250N
RCRV010410500N
Ruthenium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Ruthenium Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml