Manganese Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010395-250N RV010395-500N RV010395-50N RV010465-100N RV010395-100N RV010465-250N RV010465-50N RV010465-500N
RCRV010395500NEA 449.77 USD
RCRV010395500N RCRV010395-50N RCRV010465100N RCRV010465500N RCRV010395100N RCRV010465250N RCRV010395250N RCRV010465-50N
Manganese Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Manganese Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml