Gallium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010382-100N RV010382-500N RV010382-250N
RCRV010382500NEA 661.69 USD
RCRV010382500N RCRV010382250N RCRV010382100N
Gallium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Gallium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml