Wright's stain, Camco® Quik Stain®

Supplier: CAMBRIDGE DIAGNOSTIC PRODUCT

Synonyms: Eosin Methylene blue solution according to WrightWright's eosin methylene blue

210 210
71130-003EA 172.54 USD
71130-003 CA71130-003
Wright's stain, Camco® Quik Stain®
Wright's stain

The Fastest and Most Reliable Differential Stain Available


  • Fast: Requires less than 30 seconds
  • Reliable: Uniformity from slide to slide
  • Economical: Stains hundreds of smears
  • Simple to Use: Only one staining solution necessary


Camco Quik Stain revolutionized differential staining. One easy to use solution is all that is required for manual staining techniques. Both products incorporate a buffer into the stain solution eliminating many additional steps. Provides quick differential staining of blood smears in just 30 seconds. These products are great for a backup system in your lab or when a STAT result is necessary. These stains should be used with distilled water (pH 6-7) as the rinse.


Directions:
Prepare blood smear on slide.
Dip slide in stain for 10 seconds.
Dip slide in distilled water (pH 6-7) for 20 seconds (or more for darker staining).
Air dry.


Caution: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm. For more information, go to P65Warnings.ca.gov.

MDL Number: MFCD00082143
CAS Number: 68988-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR