Eosin Y (yellowish) solution Type V stain for histology
Supplier: CANCER DIAGNOSTICS, INC.
|
Danger
|
Synonyms:
Tetrabromofluorescein disodium salt
, Eosin Y disodium salt
, Eosin 225
, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt
, Solvent Red 43
, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate
, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Field's Stain B
, 2,4,5,7-Tetrabromofluorescein disodium salt
Eosin Type V is a class formulation of the alcoholic eosin-Y counterstain.
Eosin Type V produces brilliant counterstaining and differentiation of cell cytoplasms and other non-nuclear tissue components. Eosin Type V is ready to use and has been balanced to the proper PH for consistency and stability.
Formula:
C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 UN: 1170 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...