Petroleum spirit, 30…40 °C, Puriss. p.a., low boiling point hydrogen treated naphtha, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

77399-1L 77399-2.5L 77399-25L
BJ77399-1LEA 113.07 USD
BJ77399-1L BJ77399-2.5L BJ77399-25L
Petroleum spirit, 30…40 °C, Puriss. p.a., low boiling point hydrogen treated naphtha, Riedel-de Haën™
Petroleum spirit

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


UNSPSC Code: 12191502

Boiling Pt: 7.8 %(V)
Density: 0.628 g/cm³ (15 °C)
Flash Pt: –56 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 309-852-0
UN: 1268
ADR: 3,I

Order Now

Specification Test Results

Boiling range (30-40°C) Min. 95 %
Boiling range 30 - 40 °C
Density (D 20/4) 0.624 - 0.627
Refractive index (n 20/D) 1.356 - 1.358
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.005 %
Aluminium (Al) Max. 0.00005 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Iodine value Max. 2
Reaction against H₂SO₄ complying
Benzene (GC) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR