Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
34978-4X4L
BJ34978-4X4LCS
725.03
USD
BJ34978-4X4L
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Water with 0,1% (v/v) trifluoroacetic acid
RTECS Number: 231-791-2
Boiling Pt:
100 °C Melting Pt: 0 °C Density: 0.9982 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 76-05-1 EINECS: 200-929-3 |
Specification Test Results
Assay trifluoroacetic acid | 0.093 - 0.107 % |
Assay trifluoroacetic acid | 0.093 - 0.107 % |
pH (20 °C) | 2.0 - 2.2 |
pH (20 °C) | 2.0 - 2.2 |
Calcium (Ca) | max. 0.5 ppm |
Calcium (Ca) | max. 0.5 ppm |
Potassium (K) | max. 0.5 ppm |
Potassium (K) | max. 0.5 ppm |
Magnesium (Mg) | max. 0.5 ppm |
Magnesium (Mg) | max. 0.5 ppm |
Sodium (Na) | max. 2 ppm |
Sodium (Na) | max. 2 ppm |
Transmittance at 210 nm | min. 25 % |
Transmittance at 210 nm | min. 25 % |
Transmittance at 230 nm | min. 85 % |
Transmittance at 230 nm | min. 85 % |
Transmittance from 254 nm | min. 99 % |
Transmittance from 254 nm | min. 99 % |
HPLC Gradient at 210 nm | max. 50 mAU |
HPLC Gradient at 210 nm | max. 50 mAU |
HPLC Gradient at 254 nm | max. 10 mAU |
HPLC Gradient at 254 nm | max. 10 mAU |
LC-MS application | complying |
LC-MS application | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...