Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Methane dichloride, Methylene chloride, Methylene chloride
34897-1L
34897-6X1L
BJ34897-1LEA
126.54
USD
BJ34897-1L
BJ34897-6X1L
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Dichloromethane
- Solvents designed for high-performance thin-layer chromatography
Beilstein Number: 1730800
Formula:
CH₂Cl₂ Boiling Pt: 39…41 °C (1.013 hPa) Melting Pt: –97 °C Density: 1.32…1.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III |
|
Specification Test Results
Explosion Limit | 22 % |
Assay (GC) | Min. 99.8 % |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.02 % |
Free acid (as HCl) | Max. 0.001 % |
Transmittance at 235 nm | Min. 20 % |
Transmittance at 240 nm | Min. 50 % |
Transmittance at 245 nm | Min. 80 % |
Transmittance from 260 nm | Min. 98 % |
for AMD technic | complying |
Refractive Index | n20/D 1.424 (lit.) |
Autoignition Temperature | 1223 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...