Petroleum spirit, 50…70 °C, Puriss. p.a. Reag. Ph. Eur., high boiling, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

32247-1L 32247-2.5L
BJ32247-1LEA 120.78 USD
BJ32247-1L BJ32247-2.5L
Petroleum spirit, 50…70 °C, Puriss. p.a. Reag. Ph. Eur., high boiling, Riedel-de Haën™
Petroleum spirit

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Boiling Pt: 50…70 °C (1.013 hPa)
Density: 0.664 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 232-453-7
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Boiling range 50 - 70 °C
Density (D 20/20) 0.661 - 0.664
Refractive index (n 20/D) 1.372 - 1.376
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.002 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Benzene (GC) Max. 0.01 %
S-compounds (as S) Max. 0.005 %
Iodine value Max. 0.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR