Petroleum spirit, 40…60 °C 75, Puriss., meetSpec_DAB, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

24587-1L 24587-2.5L 24587-25L 24587-4X2.5L 24587-5L
BJ24587-1LEA 88.37 USD
BJ24587-1L BJ24587-2.5L BJ24587-25L BJ24587-4X2.5L BJ24587-5L
Petroleum spirit, 40…60 °C 75, Puriss., meetSpec_DAB, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™
Petroleum spirit

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


UNSPSC Code: 12191502

Boiling Pt: 40…60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Boiling range 40-60 °C Min. 75 %
Density (D 20/20) 0.642 - 0.656
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Benzene (GC) Max. 0.001 %
S-compounds (as S) Max. 0.005 %
Appearance complying
Acidity or alkalinity complying
n-Hexane (GC) Max. 2 %
Reaction against H₂SO₄ complying
Tetraethyl lead complying
S-compounds, reducing matters complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR