n-Pentane 95,0 GC_ASSAY_METHOD, Laboratory Reagent
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Pentane
15607-4X5L
15607-5L
BJ15607-4X5LCS
831.23
USD
BJ15607-4X5L
BJ15607-5L
n-Pentane 95,0 GC_ASSAY_METHOD, Laboratory Reagent
n-Pentane
- ≥95% for general laboratory use.
Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36…37 °C Melting Pt: -130 Density: 0.631 g/cm³ (15 °C) Flash Pt: <–35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II Merck Index: 13,07193 |
|
Specification Test Results
Assay (GC) | Min. 95.0 % |
Water (Karl Fischer) | Max. 0.02 % |
Non-volatile matter | Max. 0.002 % |
Autoignition Temperature | 500 °F |
Refractive Index | n20/D 1.358 (lit.) |
Vapor Density | 2.48 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...