Ethylbenzene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Phenylethane

03080-1L 03080-250ML
BJ03080-1LEA 221.42 USD
BJ03080-1L BJ03080-250ML
Ethylbenzene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Ethylbenzene

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1901871
RTECS Number: DA0700000
UNSPSC Code: 12190000

Formula: C₈H₁₀
Boiling Pt: 135…137 °C (1.013 hPa)
Melting Pt: –95 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 810 °F
Identity (NMR) complying
Identity (IR) complying
Assay (GC) Min. 99.0 %
Toluene (GC) Max. 0.3 %
Benzene (GC) Max. 0.1 %
Refractive index (n 20/D) 1.494 - 1.496
Non-volatile matter Max. 0.002 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Explosion Limit 6.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR