igScript™ First Strand cDNA Synthesis Kit, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
igScript™ first strand cDNA synthesis kit contains all the necessary components to generate cDNA from an RNA template.
- Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA
- Active at temperatures up to 55 °C
- Useful for first strand cDNA synthesis for PCR or RT-PCR and gene expression data validation
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 5x igScript™ master mix
- Oligo d(T)23 VN primer (50 µM)
- Random hexamer primer mix (60 µM)
- Nuclease free water
igScript™ first strand cDNA synthesis kit includes 5X igScript™ master mix which contains igScript™ reverse transcriptase, recombinant RNase inhibitor, dNTPs, an optimized buffer, MgCl₂ and protein stabilizers. igScript™ reverse transcriptase is a recombinant MMLV reverse transcriptase with reduced RNase H activity and increased thermostability. The kit also provides two optimized primers and nuclease-free water. An anchored oligo-dT primer forces the primer to anneal to the beginning of the polyA tail and the random hexamer primer mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs. The kit is highly efficient at producing full-length cDNA from long RNA templates at temperatures between 42 to 55 °C
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.