SYBR® Green I 10000x in DMSO nucleic acid gel stain

Supplier: Biotium

Warning

Synonyms: Thiazole Green

40086-0.5ML 40086-1ML
76221-740EA 210.9 USD
76221-740 76221-742
SYBR® Green I 10000x in DMSO nucleic acid gel stain
SYBR® Green I

Thiazole green, which is structurally identical to SYBR® green I nucleic acid gel stain, is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose, polyacrylamide gels and qPCR.


  • Color & Form Orange solution
  • Ex/Em 497/520nm (with DNA)


Thiazole green can also be used to detect ssDNA and RNA in denaturing agarose/formaldehyde and polyacrylamide/urea gels without any prewashing steps, although the sensitivity is lower. Thiazole green has the identical spectral properties as SYBR® green I (Ex/Em 497/520 nm with DNA).


SYBR is a registered trademark of Thermo Fisher Scientific.

Formula: C₃₂H₃₇N₄S
MW: 509.73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00284715
CAS Number: 163795-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR