ESD Badge Holders, Menda
Supplier: Desco
35012 35014 35010 35013 35011 35015
89126-980EA 3.35 USD
89126-980 89126-984 89126-976 89126-982 89126-978 89126-986
ESD Badge Holders, Menda
Personal Identification ID Card Holders
ESD badge holders are ideal for use by electronic manufacturers working with ESD sensitive components.
  • Highly visible yellow header and ESD symbol allows immediate identification of ESD protection
  • Tribocharging: Less than 100 volts
  • Available in horizontal and vertical formats
Badge holders feature an inner anti-print, non-ESD vinyl liner which will prevent ink transfer from the document to the document holder. Manufactured with a custom dissipative material made exclusively for Desco Industries and not available anywhere else.
Certifications: Dissipative Rtt: 10⁶ to 10¹⁰ ohms per ANSI/ESD S11.11