ESD Shop Travelers, Desco Industries
Supplier: Desco
34457 34454 34455 34456
10147-636PK 94.29 USD
10147-636 10147-656 10147-658 10147-766
ESD Shop Travelers, Desco Industries
Document Holders
ESD shop travelers are ideal for use by electronic manufacturers who work with ESD sensitive components.
  • Choice of four header colors to help identify group, department, and work instructions
  • Designed to hold 'A' size documents (8.5×11")
  • Reinforced hang hole
  • Silicone-free
Choice of four header colors to help keep track of everyone within a facility, or just assist in organizing meetings and events. Dissipative vinyl is 1×10⁴ to <1×10¹¹ ohms, per ANSI/ESD STM11.11. Suitable for cleanroom applications or where contamination could be a concern.