Condensers, West, Chemglass
Supplier: Chemglass
CG-1220-01 CG-1220-03 CG-1220-05 CG-1220-20 CG-1220-21 CG-1220-23 CG-1220-24
80067-404EA 198.31 USD
80067-404 80067-408 80067-412 80067-420 80067-422 80067-424 80067-426
Condensers, West, Chemglass
Condensers
Condensers feature increased flow rate of water.
  • Narrow annular space between jacket and inner tube
  • West condenser provides for excellent cooling efficiency
Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O. Ø. of 10 mm at the middle serration.