Eosin Y (yellowish), dry powder stain
Supplier: Epredia
|
Warning
|
Synonyms:
Tetrabromofluorescein disodium salt
, Eosin Y disodium salt
, Eosin 225
, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt
, Solvent Red 43
, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate
, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Field's Stain B
, 2,4,5,7-Tetrabromofluorescein disodium salt
Stain specimens with Eosin-Y compounds with Epredia™ Dry Powder Stains, certified by the Biological Stain Commission.
Enhances Differentiation Between Muscle, Red Blood Cells, and Connective Tissue
Formula:
C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...