Shandon™ OG 6 stain for cytology
Supplier: Epredia
6766005 6766006
89370-824CS 163.37 USD
89370-824 89370-826
Shandon™ OG 6 stain for cytology
Papanicolaou's Orange G solution (OG 6)
Obtain maximum performance and maximum value with Epredia™ Shandon™ OG-6 Stain.
Demonstrates keratin in cytoplasm.
IVDLogo