Jenner's stain

Supplier: Epredia

Synonyms: Eosin−Methylene Blue

89030 89030
84000-194EA 110.46 USD
84000-194 CA84000-194
Jenner's stain
Jenner's stain

Achieve results similar to that of a Giemsa stain with Epredia™ Jenner Stain.

MDL Number: MFCD00081733
CAS Number: 62851-42-7
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR