Flasks, with Drain Valves, Round Bottom, 3-Necks, Chemglass
Supplier: Chemglass
CG-1523-V-03 CG-1523-V-04 CG-1523-V-05 CG-1523-V-01 CG-1523-V-02 CG-1523-V-06
10034-228EA 1120.86 USD
10034-228 10034-230 10034-232 10034-310 10034-312 10034-350
Flasks, with Drain Valves, Round Bottom, 3-Necks, Chemglass
Flasks Round Bottom Flasks
Heavy wall round bottom three neck flasks with standard taper outer joints on center and side necks.
  • Once opened the contents of the flask will drain through the sidearm
  • The sidearm on the drain valve is 1/2" Swagelok®
Lower outlet is a drain valve extended tip which fills the seat area and permits total agitation of the contents of the flask.