Filter Tubes, Allihn, Chemglass
Supplier: Chemglass
CG-1410-01 CG-1410-02 CG-1410-03
80068-584EA 97.47 USD
80068-584 80068-586 80068-588
Filter Tubes, Allihn, Chemglass
Filters Filter Tubes
Allihn type filter tube with a 30 mm Ext.Ø disc.
  • High-quality filter feature tubes allihn and a fritted design
  • Frit choices are listed and consists of coarse, medium and fine frit
Height above the disc is 100mm, lower stem is 8×55 mm Ext.Ø in length.