Condensers, Micro Cold Finger, Chemglass
Supplier: Chemglass
CG-1222-A-01 CG-1222-A-02
80067-442EA 316.77 USD
80067-442 80067-444
Condensers, Micro Cold Finger, Chemglass
Condensers
Condenser for use in refluxing from 1 to 20 ml of solvent in flasks having a capacity of 5 to 50 ml.
  • Minimal surface area provides for better yields while providing adequate cooling
  • The additional hose connection is provided to allow introduction of an inert gas such as nitrogen
Top hose connection is cooling inlet and middle hose connection is cooling outlet. Cold finger extends either 5 or 13 mm below the joint and is used on a flask with the listed joint size. Hose connection is 10 mm at the largest serration.