Block Heater, 120 V
Supplier: DLAB SCIENTIFIC INC.
5032101200 18900221 18900222 18900223 18900224 18900274 18900275 18900276 18900277 18900279 18900278
470319-530EA 437.5 USD
470319-530 470319-532 470319-534 470319-536 470319-538 470319-540 470319-542 470319-864 470319-866 470319-868 470319-870
Block Heater, 120 V
Block Heaters Block Heater/Coolers
Affordable dry block heater for the classroom or lab.
  • 110 - 120 V, 50/60 Hz
  • Temperature control up to 120 °C (in 1 °C increments)
  • Various types of optional blocks are available
  • Overheating protection
  • Lid for protection and heat preservation
This single block heater comes with a unique aluminum block to accommodate 18 tubes of each of the following standard-sized tubes: 0.2 ml, 0.5 ml and 1.5/2 ml. The unit heats up quickly (maximum of 5.5 °C/minute), and the LED screen clearly displays the temperature and timer.