Orbital Shakers, Orbi-Shaker™ Series
Supplier: Ward's
BT1011 BT1011-E BT3001 BT3001-E BT302-E
470318-948EA 4177.9 USD
470318-948 470318-950 470318-954 470318-956 470318-960
Orbital Shakers, Orbi-Shaker™ Series
Shakers and Mixers Orbital Shakers
Compact, but powerful orbital shakers.
  • Horizontally circular orbit for aeration and mixing
  • Optional platform available for use with MAGic Clamp™ system
  • 2 year manufacturer's warranty
The powerful brushless induction drive used on this line is designed for years of trouble-free service, even under continuous use conditions with heavy loads. Noise and vibration are minimized by an advanced, computer designed motor isolation system.
The Orbi-Shakers come standard with a non-slip, rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays. All units are compatible with the optional MAGic Clamp System which allows use of the MAGic Clamp™ attachment line, for quick-changes between flask clamps of different sizes.
Ordering information: MAGic clamp platform and other accessories must be ordered separately.
Delivery information: Supplied with a non-slip, rubber mat.