Rubbermaid® Commercial ProSave™ Bin Replacement Lid and Scoop, Essendant
Supplier: Essendant
Rubbermaid®
RCP9F77
19759-120EA 613.7 USD
19759-120
Rubbermaid® Commercial ProSave™ Bin Replacement Lid and Scoop, Essendant
Foodservice Supplies
The Rubbermaid® Commercial ProSave™ Food Storage Lid with Scoop is designed to for use with Rubbermaid® Commercial ProSave™ bulk food storage containers.
  • Hook system for sanitary storage of the scoop inside of the food storage containers
  • Clear window on the lid, it is easy to see contents of the bulk food storage containers
  • Ideal for commercial garden centers and food service centers
  • Constructed from commercial grade structural web plastic
With a hook system that allows the scoop to be easily stored inside, this food storage lid works in commercial food service and garden centers. Constructed from durable industrial grade structural web plastic, this lid is easy to clean and keep sanitary.