DC Power Supply 20 V, 5 A
Supplier: AMERICAN 3B SCIENTIFIC SE
1003311
470316-728EA 398.79 USD
470316-728
DC Power Supply 20 V, 5 A
Power Supplies
Universal power supply with digital current and voltage display. Output voltage and output current are continuously adjustable.
  • DC output 0 - 20 V, 0 - 5 A
  • 100 W output power
  • 2×3 digit LED display
The device can be used as a constant voltage source with current limiting or as a constant current source with voltage limiting.
Terminals: 4 mm "banana" safety sockets
Weight: 4.7 kg