Extra Low Voltage Safety Leads
Supplier: AMERICAN 3B SCIENTIFIC SE
1002849
470316-712EA 26.1 USD
470316-712
Extra Low Voltage Safety Leads
Power Supplies
Set of two copper leads with highly flexible PVC insulation, 75 cm long, black, with stackable 4 mm safety laminated plugs at both ends.
  • Conductor Cross-Section: 2.5 mm2
  • Voltage: Safety Extra Low Voltage
  • Maximum Continuous Current: 32 A