DC Power Supply
Supplier: AMERICAN 3B SCIENTIFIC SE
1012857
470316-706EA 586.59 USD
470316-706
DC Power Supply
Power Supplies
High-quality switched-mode power supply in space-saving housing with intelligent control of fan speed to ensure safe and quiet operation.
  • DC output 1 – 32 V, 0 - 20 A (0 – 5 A output with 4-mm safety sockets on front)
  • 640 W max. output power
  • 3-digit, 15 mm, green LED display
Simple, precise and fast adjustment of voltage and current levels with dual-function rotary knobs for coarse and fine adjustment. Adjustable current limiting in open circuit. Three user-definable stored configurations for voltage and current limiting make it easy to recall frequently used settings. Full remote control of voltage and current plus output which can be turned on and off.
Dimensions: 200×90×255 mm
Weight: 2.6 kg