P10035 Digital Multimeter
Supplier: AMERICAN 3B SCIENTIFIC SE
1002781
470316-702EA 61.19 USD
470316-702
P10035 Digital Multimeter
Electrical Meters Digital Multimeters
Compact 31/2" digit multimeter for measuring voltage, current and resistance and also including diode and continuity tests. Complete with pouch, leads and battery.