Eosin Y 1% in aqueous solution stain for histology

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Acid Red 872,4,5,7-TetrabromofluoresceinEosin yellowishSolvent red 43BromofluoresceinSpiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-CI 45380Tetrabromofluorescein2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acidBronze Bromo ESBromo acid J. TS, XL, or XX

26763-01
101411-526EA 57.97 USD
101411-526
Eosin Y 1% in aqueous solution stain for histology
Eosin Y
Formula: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
MDL Number: MFCD00036189
CAS Number: 15086-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR