Cardionics® Bionic Hybrid System
Supplier: AMERICAN 3B SCIENTIFIC SE
1022278 1022279 1022273 1022455 1021981 1022280 1022454 1022282 1022452 1022453 1022275 1022281
470318-040EA 6001.09 USD
470318-040 470318-030 470318-042 470318-032 470318-026 470318-028 470318-022 470318-044 470318-012 470318-034 470318-046 470318-024
Cardionics® Bionic Hybrid System
Models Human Biology Models
Learn Auscultation with a living patient.
  • Use with a simulator, manikin, or standardized patient
  • Ausculate, Record EKG, Measure Pulse
  • Washable
When worn by the standardized patient, the suit simulates physiological conditions to test diagnostic and procedural skills. It is controlled wirelessly with easy to use software and responds in real time to diagnosis and treatment with direct feedback.
Features 5-wire EKG connections, pulse points, optional blood pressure cuff accessory, as well as auscultation capabilities. Instructors can easily adjust and adapt the suit scenarios to fit many conditions wirelessly by using the SimScope™ Wi-Fi.
SimScope™ Wi-Fi and tablet included with system. Reusable and washable (30 °C), available in different sizes.
For Auscultation practice only, use a SimShirt. Similar to the Bionic Hybrid. The SimShirt uses the same technology for Auscultation points, but doesn't have the Pulse, blood pressure or EKG connections.