Cardionics® SAM II Student Auscultation Manikin
Supplier: AMERICAN 3B SCIENTIFIC SE
1022656 1021555 1020095
470318-050EA 7200.09 USD
470318-050 470317-920 470317-954
Cardionics® SAM II Student Auscultation Manikin
Models Human Biology Models
Extended Auscultation Capabilities!
  • Includes COVID sounds
  • Lightweight and portable
  • Includes lesson guide
  • Use with any stethoscope
  • Palpable carotid pulse
Auscultation is an essential clinical skill needed to assess and monitor patients’ conditions. With the Cardionics SAM II Student Auscultation Manikin, teaching and learning this skill becomes even more versatile and economic. The SAM II trainer can be used with any stethoscope. All sounds and videos are recorded from live patients for students to experience a life-like simulation. Users can also create and save their own case videos.
SAM II Student Auscultation Manikin is used in teaching and learning heart, lung and bowel sounds. When connected to the laptop (included) with the pre-installed software, a variety of sounds, videos and lessons recorded can be accessed.
Additionally, when connected to speakers SAM II Student Auscultation Manikin can be easily moved into a classroom or an auditorium for group instruction, where a larger audience of students can benefit from a co-learning experience and share their thoughts at the same time as high quality heart, lung and bowel sounds are emitted. The programmable and password protected lectures allow teaching institutions to have numerous instructors utilize the SAM II trainer. Included with the product is SAM’s Lesson Guide: a complete guide of lessons for each auscultation sound that SAM II offers.
If you already have a SAM II, or want to upgrade in the future, a software upgrade package is available.