Nylon Biopsy Bags, Simport Scientific
Supplier: Simport
M478-3 M478-1
10154-076PK 541.42 USD
10154-076 10154-038
Nylon Biopsy Bags, Simport Scientific
Bags Sample Bags
A safe and convenient way to protect your sample during processing.
  • Made of nylon
  • Solvent resistant
  • Excellent fluid exchange
  • Maximum tissue safety with see through bag
These sturdy biopsy bags are made of white nylon thin mesh (0.3 mm Ø), reducing the risk of specimen loss during processing.  While solvent resistant, they provide great tissue safety and excellent fluid exchange.  In order to ease specimen removal, bags can be peeled open.  All seams are heat-sealed.