Nitric acid 68-70% ACS

Supplier: AQUA SOLUTIONS MS
Danger

Synonyms: Nitric acid solution

N3805-500ML N3805-2.5L
101204-214EA 112.59 USD
101204-214 101203-496
Nitric acid 68-70% ACS
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8 (5.1) II
Merck Index: 13,06610

Order Now

Specification Test Results

Assay 68.0 – 70.0 w/w %
Aluminum ≤ 0.2 ppm
Arsenic (As) ≤ 0.004 ppm
Boron (B) ≤ 0.1 ppm
Calcium (Ca) ≤ 0.2 ppm
Chloride ≤ 0.08 ppm
Chromium (Cr) ≤ 0.1 ppm
Color ≤ 10 APHA
Copper (Cu) ≤ 0.05 ppm
Gold (Au) ≤ 0.3 ppm
Heavy Metals (as Pb) ≤ 0.2 ppm
Iron (Fe) ≤ 0.2 ppm
Lead (Pb) ≤ 0.1 ppm
Magnesium (Mg) ≤ 0.3 ppm
Nickel (Ni) ≤ 0.05 ppm
Phosphate (PO4) ≤ 0.2 ppm
Potassium (K) ≤ 0.3 ppm
Residue After Ignition ≤ 2 ppm
Sodium (Na) ≤ 0.3 ppm
Sulfate (SO4) ≤ 0.5 ppm
Tin (Sn) ≤ 0.3 ppm
Titanium (Ti) ≤ 0.3 ppm
Zinc (Zn) ≤ 0.3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR